Learn More
Cayman Chemical Sphingosine (d17:1), Purity: ≥98%, CAS: 6918-48-5, 25 mg
SDP
Catalog No. 502889976
Encompass_Preferred
A naturally-occurring but uncommon form of sphingosine, accounting for approximately 13% of the sphingosine in human skin; can be phosphorylated by sphingosine kinases to produce C17 sphingosine-1-phosphate
Catalog No. 50-288-9976 Supplier Cayman Chemical Supplier No. 1000790225 MG
A naturally-occurring but uncommon form of sphingosine, accounting for approximately 13% of the sphingosine in human skin; can be phosphorylated by sphingosine kinases to produce C17 sphingosine-1-phosphate