Learn More
Cayman Chemical Sphingosine d18 1
SDP
Catalog No. 502890135
Encompass_Preferred
Sphingosine (d18:1) is formed primarily from the breakdown of ceramide. Sphingosine inhibits PKC and phosphatidic acid phosphohydrolase, whereas it activates PLD and DAG kinase. Phosphorylation of sphingosine by SPHK 1 and SPHK 2 produces sphingosine-1-phosphate, a potent bioactive lipid that exhibits a broad spectrum of biological activities including cell proliferation, survival, migration, cytoskeletal organization, and morphogenesis.
Catalog No. 50-289-0135 Supplier Cayman Chemical Supplier No. 1000790750 MG
Add to Cart
Add to Cart
Sphingosine (d18:1) is formed primarily from the breakdown of ceramide. Sphingosine inhibits PKC and phosphatidic acid phosphohydrolase, whereas it activates PLD and DAG kinase. Phosphorylation of sphingosine by SPHK 1 and SPHK 2 produces sphingosine-1-phosphate, a potent bioactive lipid that exhibits a broad spectrum of biological activities including cell proliferation, survival, migration, cytoskeletal organization, and morphogenesis.