Learn More
Cayman Chemical Sphingosine (d20:1), Purity: ≥98%, CAS: 6918-49-6, 10 mg
SDP
Catalog No. 502890109
Encompass_Preferred
Composes a significant portion of CNS gangliosides; concentration increases with maturation and may be related to nervous system degeneration
Catalog No. 50-289-0109 Supplier Cayman Chemical Supplier No. 1000790310 MG
Composes a significant portion of CNS gangliosides; concentration increases with maturation and may be related to nervous system degeneration