Learn More
Cayman Chemical Sphingosine d20 1
SDP
Catalog No. 502890993
Encompass_Preferred
Composes a significant portion of CNS gangliosides; concentration increases with maturation and may be related to nervous system degeneration
Catalog No. 50-289-0993 Supplier Cayman Chemical Supplier No. 10007903500 UG
Add to Cart
Add to Cart
Composes a significant portion of CNS gangliosides; concentration increases with maturation and may be related to nervous system degeneration