Learn More
Bioss Sphingosine Kinase 1 C-termin
SDP
Catalog No. 5000065831
Encompass_Preferred
Sphingosine Kinase 1 (C-terminal region) Primary Antibody Unconjugated Host Species Rat Target Species Human Mouse Rat Clonality Monoclonal Applications WB M540Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65831 Supplier Bioss Supplier No. BSM70675T
Sphingosine Kinase 1 (C-terminal region) Primary Antibody Unconjugated Host Species Rat Target Species Human Mouse Rat Clonality Monoclonal Applications WB M540Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol