Learn More
Cayman Chemical Sphingosine Kinase Inhibitor 2, Purity: ≥95%, CAS: 312636-16-1, 50 mg
SDP
Catalog No. 502890910
Encompass_Preferred
An SPHK1 inhibitor (IC50 = 0.5 µM for non-ATP-competitive inhibition of human recombinant SPHK1); selective for SPHK1 over ERK2, PI3K, and PKCα at 50s = 0.9-4.6 µM)
Catalog No. 50-289-0910 Supplier Cayman Chemical Supplier No. 1000922250 MG
An SPHK1 inhibitor (IC50 = 0.5 µM for non-ATP-competitive inhibition of human recombinant SPHK1); selective for SPHK1 over ERK2, PI3K, and PKCα at 50s = 0.9-4.6 µM)