Learn More
Echelon Biosciences Research Labs SPHINGOSINE-TMR 50 ug
SDP
Supplier:  Echelon Biosciences Research Labs S100T50UG
Encompass
Sphingosine-tetramethylrhodamine is an alkyl-labeled sphingosine synthesized with the natural erythro stereochemistry that maintains its natural properties.Echelon offers a unique selection of sphingosine analogs labeled at the terminus of the alkyl chain with biotin or fluorescent groups. All compounds are synthesized from chiral precursors and are available as lyophilized powders. Echelons alkyl-labeled sphingosines are synthesized with the natural erythro stereochemistry and maintain similar properties as sphingosine. For example, labeled sphingosines are phosphorylated by sphingosine kinases (1 & 2) and can be used in high throughput screening applications. In addition, they induce a Nieman Pick type C1 disease cellular phenoype in RAW macrophages in the same manner as exogenously added sphingosine. This item is non returnable
Catalog No. 50-720-766