Learn More
Cayman Chemical Sphingosine1phosphated7 d18
SDP
Catalog No. 502887754
Encompass_Preferred
An internal standard for the quantification of S1P by GC- and LC-MS
Catalog No. 50-288-7754 Supplier Cayman Chemical Supplier No. 10008121500 UG
An internal standard for the quantification of S1P by GC- and LC-MS