Learn More
Echelon Biosciences Research Labs SPHINGOSTRIPS 2 pack
SDP
Catalog No. 50720777
Encompass
Sphingo Strips are 2 x 6 cm hydrophobic membranes that have been spotted with 100 pmol of fifteen different sphingolipids. They are used to determine how your protein of interest interacts with one or more of these lipids in a simple protein lipid overlay assay. Bound protein is then detected by an antibody against the protein or affinity tag. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-777 Supplier Echelon Biosciences Research Labs Supplier No. S60002PK
Sphingo Strips are 2 x 6 cm hydrophobic membranes that have been spotted with 100 pmol of fifteen different sphingolipids. They are used to determine how your protein of interest interacts with one or more of these lipids in a simple protein lipid overlay assay. Bound protein is then detected by an antibody against the protein or affinity tag. This item is non returnable