Learn More
Echelon Biosciences Research Labs SPHINGOBEAD SAMPLE PACK
SDP
Supplier:  Echelon Biosciences Research Labs PB00SSEAPK
Encompass
Sample pack of 9 lipid coated beads. Each order of beads includes a small amount of control beads. Each vial contains 0.1mL agarose beads (50% slurry in PBS buffer) with 1 nanomole of bound lipid. Larger quantities of control beads are also available for purchase. This item is non returnable
Catalog No. 50-720-684