Learn More
Selleck Chemical LLC SPHINX31 5mg 1818389-84-2
SDP
Supplier:  Selleck Chemical LLC S88105MG
Encompass_Preferred
SPHINX31 is a SRPK1 inhibiror with an IC50 of 5.9 nM with highly selectivity for SRPK1 over SRPK2 (50-fold) and CLK1 (100-fold). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8436