Learn More
Cayman Chemical SPHK Substrate 1ea
SDP
Catalog No. 502896692
Encompass_Preferred
A PD-1/PD-L1 interaction inhibitor (IC50 6.3 nM) increases T cell proliferation in a PD-L1-overexpressing CT26 colon cancer cell and T cell co-culture at 20 nM promotes T cell infiltration into an in vitro PD-L1-overexpressing murine Lewis lung carcinoma tumor matrix at 20 and 80 nM reduces tumor growth and promotes CD8 and CD4 T cell tumor infiltration in a PD-L1-overexpressing murine Lewis lung carcinoma model at 20 mg/kg
Catalog No. 50-289-6692 Supplier Cayman Chemical Supplier No. 7017431 EA
A PD-1/PD-L1 interaction inhibitor (IC50 6.3 nM) increases T cell proliferation in a PD-L1-overexpressing CT26 colon cancer cell and T cell co-culture at 20 nM promotes T cell infiltration into an in vitro PD-L1-overexpressing murine Lewis lung carcinoma tumor matrix at 20 and 80 nM reduces tumor growth and promotes CD8 and CD4 T cell tumor infiltration in a PD-L1-overexpressing murine Lewis lung carcinoma model at 20 mg/kg