Learn More
Cayman Chemical Sphinganine-1-phosphate (d17:0), Purity: ≥95%, CAS: 474923-29-0, 10 mg
SDP
Catalog No. 502987726
Encompass_Preferred
A sphingolipid; levels are lower in human trabecular meshwork samples from patients with glaucoma compared with control samples
Catalog No. 50-298-7726 Supplier Cayman Chemical Supplier No. 2250110 MG
A sphingolipid; levels are lower in human trabecular meshwork samples from patients with glaucoma compared with control samples