Learn More
Cayman Chemical SphngomyelInbovIn spInal 1g
SDP
Catalog No. 502994867
Encompass_Preferred
A mixture of bioactive sphingolipids; found in mammalian cell membranes; make up 2-15% of the total organ phospholipid population but are found at higher concentrations in the brain and myelin sheaths surrounding peripheral nerves; interact with cholesterol to control its distribution within cellular membranes and maintain cholesterol homeostasis in cells; undergo hydrolysis by sphingomyelinase to form ceramides
Catalog No. 50-299-4867 Supplier Cayman Chemical Supplier No. 226741 G
A mixture of bioactive sphingolipids; found in mammalian cell membranes; make up 2-15% of the total organ phospholipid population but are found at higher concentrations in the brain and myelin sheaths surrounding peripheral nerves; interact with cholesterol to control its distribution within cellular membranes and maintain cholesterol homeostasis in cells; undergo hydrolysis by sphingomyelinase to form ceramides