Learn More
Cayman Chemical Sphingomyelin (bovine spinal cord), Purity: ≥95% (natural bovine mixture), CAS: 85187-10-6, 25 mg
SDP
Catalog No. 502964552
Encompass_Preferred
A mixture of bioactive sphingolipids; found in mammalian cell membranes; make up 2-15% of the total organ phospholipid population but are found at higher concentrations in the brain and myelin sheaths surrounding peripheral nerves; interact with cholesterol to control its distribution within cellular membranes and maintain cholesterol homeostasis in cells; undergo hydrolysis by sphingomyelinase to form ceramides
Catalog No. 50-296-4552 Supplier Cayman Chemical Supplier No. 2267425 MG
A mixture of bioactive sphingolipids; found in mammalian cell membranes; make up 2-15% of the total organ phospholipid population but are found at higher concentrations in the brain and myelin sheaths surrounding peripheral nerves; interact with cholesterol to control its distribution within cellular membranes and maintain cholesterol homeostasis in cells; undergo hydrolysis by sphingomyelinase to form ceramides