Learn More
Cayman Chemical Sphingomyelin (chicken egg), Purity: ≥95%, CAS: 383907-87-7, 5 mg
SDP
Catalog No. 502934969
Encompass_Preferred
A mixture of bioactive sphingolipids; found in mammalian cell membranes; make up 2-15% of the total organ phospholipid population but are found at higher concentrations in the brain and myelin sheaths surrounding peripheral nerves; interact with cholesterol to control its distribution within cellular membranes and maintain cholesterol homeostasis in cells; undergo hydrolysis by sphingomyelinase to form ceramides
Catalog No. 50-293-4969 Supplier Cayman Chemical Supplier No. 243455 MG
A mixture of bioactive sphingolipids; found in mammalian cell membranes; make up 2-15% of the total organ phospholipid population but are found at higher concentrations in the brain and myelin sheaths surrounding peripheral nerves; interact with cholesterol to control its distribution within cellular membranes and maintain cholesterol homeostasis in cells; undergo hydrolysis by sphingomyelinase to form ceramides