Learn More
Cayman Chemical Sphingomyelin (porcine RBC), Purity: ≥98%, CAS: 85187-10-6, 50 mg
SDP
Catalog No. 502991872
Encompass_Preferred
A mixture of bioactive sphingolipids; found in mammalian cell membranes; make up 2-15% of the total organ phospholipid population but are found at higher concentrations in the brain and myelin sheaths surrounding peripheral nerves; interact with cholesterol to control its distribution within cellular membranes and maintain cholesterol homeostasis in cells; undergo hydrolysis by sphingomyelinase to form ceramides
Catalog No. 50-299-1872 Supplier Cayman Chemical Supplier No. 3155850 MG
A mixture of bioactive sphingolipids; found in mammalian cell membranes; make up 2-15% of the total organ phospholipid population but are found at higher concentrations in the brain and myelin sheaths surrounding peripheral nerves; interact with cholesterol to control its distribution within cellular membranes and maintain cholesterol homeostasis in cells; undergo hydrolysis by sphingomyelinase to form ceramides