Learn More
Cayman Chemical Sphingosine-1-phosphate (d17:1), Purity: ≥95%, CAS: 474923-27-8, 5 mg
SDP
Catalog No. 502987727
Encompass_Preferred
A synthetic sphingolipid; has been used as an internal standard for the quantification of S1P (d18:1) in plasma and tissue samples
Catalog No. 50-298-7727 Supplier Cayman Chemical Supplier No. 224985 MG
A synthetic sphingolipid; has been used as an internal standard for the quantification of S1P (d18:1) in plasma and tissue samples