Learn More
Cayman Chemical Sphingosine-13C2-d2 (d18:1), Purity: ≥99% deuterated forms (d1-d2), CAS: 2692623-81-5, 1 mg
SDP
Catalog No. 502940221
Encompass_Preferred
An internal standard for the quantification of sphingosine (d18:1) by GC- or LC-MS
Catalog No. 50-294-0221 Supplier Cayman Chemical Supplier No. 278751 MG
An internal standard for the quantification of sphingosine (d18:1) by GC- or LC-MS