Learn More
Cayman Chemical SphngosIn KInase 2humn25ug
SDP
Catalog No. 502990090
Encompass_Preferred
Source: Active recombinant N-terminal His-tagged protein expressed in insect cells • Amino acids: 2-654 (full length) • MW: 72.9 kDa
Catalog No. 50-299-0090 Supplier Cayman Chemical Supplier No. 3019125 UG
Source: Active recombinant N-terminal His-tagged protein expressed in insect cells • Amino acids: 2-654 (full length) • MW: 72.9 kDa