Learn More
Cayman Chemical SphngosInd14 1 10mg
SDP
Catalog No. 502935887
Encompass_Preferred
A bioactive sphingolipid; has been found in B. mori, P. clarkii, and A. aurita extracts; increases the germination rate of N. rileyi (EC50 = 0.0102 ng/ml); inhibits PKC in vitro (IC50 = 7.3 mol %); inhibits PMA-induced superoxide generation in neutrophils and reduces growth of CHO cells (IC50s = 19 and 8 μM, respectively)
Catalog No. 50-293-5887 Supplier Cayman Chemical Supplier No. 2437310 MG
A bioactive sphingolipid; has been found in B. mori, P. clarkii, and A. aurita extracts; increases the germination rate of N. rileyi (EC50 = 0.0102 ng/ml); inhibits PKC in vitro (IC50 = 7.3 mol %); inhibits PMA-induced superoxide generation in neutrophils and reduces growth of CHO cells (IC50s = 19 and 8 μM, respectively)