Learn More
Medchemexpress LLC Sphondin | 483-66-9 | 1 MG
SDP
Catalog No. 5000420931
Encompass_Preferred
Sphondin possesses an inhibitory effect on IL-1β-induced increase in the level of COX-2 protein and PGE2 release in A549 cells.
  • Inhibits COX-2.
  • Attenuates IL-1β-induced COX-2 protein expression and PGE2 release in A549 cells.
Catalog No. 50-004-20931 Supplier Medchemexpress LLC Supplier No. HYN24291MG
Sphondin possesses an inhibitory effect on IL-1β-induced increase in the level of COX-2 protein and PGE2 release in A549 cells.
  • Inhibits COX-2.
  • Attenuates IL-1β-induced COX-2 protein expression and PGE2 release in A549 cells.