Learn More
Strem, An Ascensus Company CAS# 657408-07-6. 2-Dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl, min. 98% SPhos. MFCD05861611. 10G
SDP
Catalog No. 501959538
Encompass
CAS# 657408-07-6. 10G. 2-Dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl, min. 98% SPhos. MFCD05861611. Molecular Formula: C26H35O2P. Molecular Weight: 410.53. Color/Form: white xtl. Strem#: 15-1143 www.strem.com/catalog/v3/15-1143/
Catalog No. 50-195-9538 Supplier Strem, An Ascensus Company Supplier No. 15114310G
CAS# 657408-07-6. 10G. 2-Dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl, min. 98% SPhos. MFCD05861611. Molecular Formula: C26H35O2P. Molecular Weight: 410.53. Color/Form: white xtl. Strem#: 15-1143 www.strem.com/catalog/v3/15-1143/