Learn More
Strem, An Ascensus Company CAS# 657408-07-6. 2-Dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl, min. 98% SPhos. MFCD05861611. 50G
SDP
Catalog No. 501959539
Encompass
CAS# 657408-07-6. 50G. 2-Dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl, min. 98% SPhos. MFCD05861611. Molecular Formula: C26H35O2P. Molecular Weight: 410.53. Color/Form: white xtl. Strem#: 15-1143 www.strem.com/catalog/v3/15-1143/
Catalog No. 50-195-9539 Supplier Strem, An Ascensus Company Supplier No. 15114350G
CAS# 657408-07-6. 50G. 2-Dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl, min. 98% SPhos. MFCD05861611. Molecular Formula: C26H35O2P. Molecular Weight: 410.53. Color/Form: white xtl. Strem#: 15-1143 www.strem.com/catalog/v3/15-1143/