Learn More
Strem, An Ascensus Company CAS# 1375325-64-6. 250mg. Chloro(2-dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl) palladium(II) min. 98% [SPhos Palladacycle Gen. 2]. MFCD20264897
SDP
Supplier:  Strem, An Ascensus Company 460283250MG
Encompass
CAS# 1375325-64-6. 250mg. Chloro(2-dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl) palladium(II) min. 98% [SPhos Palladacycle Gen. 2]. MFCD20264897. Molecular Weight: 720.62. Molecular Formula: C38H45ClNO2PPd. Color/form: white pwdr. Strem# 46-0283. http://www.strem.com/catalog/v/46-0283/
Catalog No. 50-201-6762