Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sphos Pd G4

Catalog No. 8042821G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-428-21G 250 mg
80-428-2100MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 80-428-21G Supplier Sigma Aldrich Supplier No. 8042821G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

SPhos Pd G4 is a versatile 4th generation palladium precatalyst developed by Buchwald group.

Specifications

CAS 1599466-85-9
Melting Point 240°C
Molecular Formula C44H58NO5PPdS
Linear Formula C44H58NO5PPdS
Quantity 250 mg
Molecular Weight (g/mol) 850.39 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.