Learn More
Electron Microscopy Sciences Lab Grade Narrow Neck Lab Dewar, 2000 mL with standard base, 255 mm O.D., 456mm height
SDP
Catalog No. 501930910
Encompass
These lab grade dewars are manufactured of borosilicate glass for liquid nitrogen service. A sturdy mesh cover facilitates a firm grip, provides a cushion against bumping and increases protection. Rated the most efficient glass dewars as a result of rigid quality control along with superior design and manufacturing features.
The weighted bases result in increased stability.
The dewars are supplied with a stopper to minimize evaporation
Evacuation tip is completely shielded for increased protection.
Efficiency is assured by rigid bakeout and evacuation to 10-5 torr or better. Ideal for temporary storage or for room-to-room transfer.
Volume: 2000 mL
Type: Std Base
O.D.: 255 mm
Height: 456mm
Catalog No. 50-193-0910 Supplier Electron Microscopy Sciences Supplier No. 6188812
These lab grade dewars are manufactured of borosilicate glass for liquid nitrogen service. A sturdy mesh cover facilitates a firm grip, provides a cushion against bumping and increases protection. Rated the most efficient glass dewars as a result of rigid quality control along with superior design and manufacturing features.
The weighted bases result in increased stability.
The dewars are supplied with a stopper to minimize evaporation
Evacuation tip is completely shielded for increased protection.
Efficiency is assured by rigid bakeout and evacuation to 10-5 torr or better. Ideal for temporary storage or for room-to-room transfer.
Volume: 2000 mL
Type: Std Base
O.D.: 255 mm
Height: 456mm