Learn More
Med Vet International Aneroid Sphygmomanometer Unit BASIC Adult Cuff Nylon 23 - 40 cm Pocket Aneroid, 01-776MCE, Each
Catalog No. 503045565
Encompass_Preferred
Cuff Size: Adult Color: Navy Blue Black enamel 300 mm Hg manometer Navy blue nylon cuff with range markings Inflation bladder and bulb Chrome-plated deflation valve Complete with leatherette zippered carrying case 5-year calibration Manufacturer Warranty; 1-year inflation system Manufacturer Warranty
Catalog No. 50-304-5565 Supplier Med Vet International Supplier No. 363779EA
Cuff Size: Adult Color: Navy Blue Black enamel 300 mm Hg manometer Navy blue nylon cuff with range markings Inflation bladder and bulb Chrome-plated deflation valve Complete with leatherette zippered carrying case 5-year calibration Manufacturer Warranty; 1-year inflation system Manufacturer Warranty