Learn More
Cayman Chemical Sphynolacton-7 10mg
SDP
Catalog No. 502939086
Encompass_Preferred
An agonist of S. hermonthica ShHTL7; binds to ShHTL7 (IC50 = 0.31 µM) over ShHTL2-6, ShHTL9-1, and the strigolactone receptor AtD14 at 10 µM but does inhibit ShHTL8 and ShHTL11 (IC50s = 1.2 and 7.8 µM, respectively); induces fatal S. hermonthica seed germination (MEC = 10 fM); induces suicidal germination of S. hermonthica seeds and decreases the number of plants emerging from the soil, as well as reduces S. hermonthica-induced senescence in maize plants when pre-applied to the soil of co-cultivated maize plants and S. hermonthica seeds at 10 nM
Catalog No. 50-293-9086 Supplier Cayman Chemical Supplier No. 2692210 MG
An agonist of S. hermonthica ShHTL7; binds to ShHTL7 (IC50 = 0.31 µM) over ShHTL2-6, ShHTL9-1, and the strigolactone receptor AtD14 at 10 µM but does inhibit ShHTL8 and ShHTL11 (IC50s = 1.2 and 7.8 µM, respectively); induces fatal S. hermonthica seed germination (MEC = 10 fM); induces suicidal germination of S. hermonthica seeds and decreases the number of plants emerging from the soil, as well as reduces S. hermonthica-induced senescence in maize plants when pre-applied to the soil of co-cultivated maize plants and S. hermonthica seeds at 10 nM