Learn More
Medchemexpress LLC Spi-112 | 1051387-90-6 | 98.0% | 468.46 | C22H17FN4O5S | 5 MG
SDP
Catalog No. 5000247990
Encompass_Preferred
SPI-112 is a small-molecule, selective and competitive inhibitor of SHP2 (PTPN11) used for biochemical and cellular research. It inhibits SHP2 and related protein tyrosine phosphatases and is supplied as a solid with guidance for storage and preparation of solution stocks.
  • Selective and competitive inhibition of SHP2 for pathway studies.
  • Reported IC50 values support biochemical assay characterization.
  • High purity solid suitable for preparation of DMSO stocks.
  • Available in small milligram quantities and as a prepared solution for convenience.
  • Storage recommendations provided to preserve compound stability.
Catalog No. 50-002-47990 Supplier Medchemexpress LLC Supplier No. HY1019645MG
SPI-112 is a small-molecule, selective and competitive inhibitor of SHP2 (PTPN11) used for biochemical and cellular research. It inhibits SHP2 and related protein tyrosine phosphatases and is supplied as a solid with guidance for storage and preparation of solution stocks.
  • Selective and competitive inhibition of SHP2 for pathway studies.
  • Reported IC50 values support biochemical assay characterization.
  • High purity solid suitable for preparation of DMSO stocks.
  • Available in small milligram quantities and as a prepared solution for convenience.
  • Storage recommendations provided to preserve compound stability.