Learn More
TARGETMOL CHEMICALS INC SPI-112 50MG
SDP
Catalog No. 502878663
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. SPI-112, the SPI-112 methyl ester analog, can inhibit cellular Shp2 PTP activity. SPI-112 bound to Shp2 by surface plasmon resonance (SPR) and displayed competitive inhibitor kinetics to Shp2. Purity 100%
Catalog No. 50-287-8663 Supplier TARGETMOL CHEMICALS INC Supplier No. T434050MG
Also available in 1 mL, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. SPI-112, the SPI-112 methyl ester analog, can inhibit cellular Shp2 PTP activity. SPI-112 bound to Shp2 by surface plasmon resonance (SPR) and displayed competitive inhibitor kinetics to Shp2. Purity 100%