Learn More
MSC SPI 4-Way V-Block, Cast Iron, 90°, 2 in. Capacity, 1 Pair
Catalog No. 5000510949
Encompass
This 4-way V-block is designed for precise machine setup, calibration, and layout tasks. Constructed from close-grained cast iron, it features a 90° V-angle with a 1/8 in. groove at the bottom, providing versatility for supporting various workpiece diameters.
  • Manufactured from durable cast iron
  • Supplied as a matched and numbered pair
  • Four distinct V-sizes support a wide diameter range from 3/4 in. to 2 in.
  • Precisely machined and ground square and parallel within 0.0008 in.
  • V-groove is parallel to the bearing surface within 0.0008 in.
Catalog No. 50-005-10949 Supplier MSC Supplier No. 88637293
This 4-way V-block is designed for precise machine setup, calibration, and layout tasks. Constructed from close-grained cast iron, it features a 90° V-angle with a 1/8 in. groove at the bottom, providing versatility for supporting various workpiece diameters.
  • Manufactured from durable cast iron
  • Supplied as a matched and numbered pair
  • Four distinct V-sizes support a wide diameter range from 3/4 in. to 2 in.
  • Precisely machined and ground square and parallel within 0.0008 in.
  • V-groove is parallel to the bearing surface within 0.0008 in.