Learn More
MSC SPI Absolute Electronic Indicator, 0.5 in. Range, 0.00016 in. Accuracy, 1 ea
Catalog No. 5000515009
Encompass
The Absolute Electronic Indicator is a high-precision digital gauge designed for accurate measurement in both inch and metric units. Featuring a 12.7 mm (0.5 in.) measurement range, this indicator is equipped for versatile use with switchable measuring modes and data output capabilities.
  • Resolution of 0.001 mm (0.00005 in.) and accuracy of 0.004 mm (0.00016 in.)
  • Equipped with a 330° rotatable LCD screen
  • Supports absolute and incremental measurement modes with SPC/USB data output
  • Includes center lug and flat back mounting options
  • Powered by a CR2032 battery and furnished in a fitted case
  • Features inch/metric conversion, tolerance functions, and measurement direction changes
  • Stem diameter of 9.5 mm (3/8 in.) with a #4-48 thread size
Catalog No. 50-005-15009 Supplier MSC Supplier No. 54325832
Add to Cart
Add to Cart
The Absolute Electronic Indicator is a high-precision digital gauge designed for accurate measurement in both inch and metric units. Featuring a 12.7 mm (0.5 in.) measurement range, this indicator is equipped for versatile use with switchable measuring modes and data output capabilities.
  • Resolution of 0.001 mm (0.00005 in.) and accuracy of 0.004 mm (0.00016 in.)
  • Equipped with a 330° rotatable LCD screen
  • Supports absolute and incremental measurement modes with SPC/USB data output
  • Includes center lug and flat back mounting options
  • Powered by a CR2032 battery and furnished in a fitted case
  • Features inch/metric conversion, tolerance functions, and measurement direction changes
  • Stem diameter of 9.5 mm (3/8 in.) with a #4-48 thread size