Learn More
MSC SPI Adjustable Angle Block, 3-3/4 in Long, 0-60° Adjustable, 50-55 RC Hardness
Catalog No. 5000530409
Encompass
This adjustable angle block is constructed from tool steel and features a hardness of 50-55 RC. It measures 3-3/4 inches long, 1-3/4 inches wide, and 2 inches high, with an adjustable angle range from 0 to 60 degrees. This block is suitable for various precision measurement and machining applications.
  • Constructed from tool steel.
  • Measures 3-3/4 inches long, 1-3/4 inches wide, and 2 inches high.
  • Offers an adjustable angle range from 0 to 60 degrees.
  • Features 50-55 RC hardness.
Catalog No. 50-005-30409 Supplier MSC Supplier No. 84533801
This adjustable angle block is constructed from tool steel and features a hardness of 50-55 RC. It measures 3-3/4 inches long, 1-3/4 inches wide, and 2 inches high, with an adjustable angle range from 0 to 60 degrees. This block is suitable for various precision measurement and machining applications.
  • Constructed from tool steel.
  • Measures 3-3/4 inches long, 1-3/4 inches wide, and 2 inches high.
  • Offers an adjustable angle range from 0 to 60 degrees.
  • Features 50-55 RC hardness.