Learn More
MSC SPI Adjustable Angle Block, Tool Steel, 0 to 60°, 50-55 RC Hardness
Catalog No. 5000527599
Encompass
This adjustable angle block is constructed from durable tool steel with a hardness of 50-55 RC. It provides a versatile solution for machining and inspection tasks, offering an adjustable angle range from 0 to 60 degrees.
  • Adjustable angle range from 0 to 60 degrees
  • Constructed from tool steel for durability
  • Hardness of 50-55 RC for precision and longevity
  • Measures 3-3/4 inch long x 1-3/16 inch wide x 2 inch high
Catalog No. 50-005-27599 Supplier MSC Supplier No. 84533819
This adjustable angle block is constructed from durable tool steel with a hardness of 50-55 RC. It provides a versatile solution for machining and inspection tasks, offering an adjustable angle range from 0 to 60 degrees.
  • Adjustable angle range from 0 to 60 degrees
  • Constructed from tool steel for durability
  • Hardness of 50-55 RC for precision and longevity
  • Measures 3-3/4 inch long x 1-3/16 inch wide x 2 inch high