Learn More
MSC SPI Analog Compression & Tension Force Gage, 11 lb/4.5 kg, 0.05 lb, 1% Accuracy
Catalog No. 5000527946
Encompass
This analog compression and tension force gage is designed for accurate measurement across a range of applications. It features a dual-capacity design, measuring up to 11 lb or 4.5 kg with a resolution of 0.05 lb and an accuracy of 1%. This versatile instrument can measure both compression and tension, making it suitable for various testing and quality control tasks.
  • Measures both compression and tension for versatile applications.
  • Provides an 11 lb/4.5 kg capacity for a wide range of measurements.
  • Offers a 0.05 lb resolution for precise readings.
  • Ensures reliable results with 1% accuracy.
  • Constructed with a durable plastic housing.
  • Includes a short form certificate of calibration, essential accessories, and a protective case.
Catalog No. 50-005-27946 Supplier MSC Supplier No. 38442075
This analog compression and tension force gage is designed for accurate measurement across a range of applications. It features a dual-capacity design, measuring up to 11 lb or 4.5 kg with a resolution of 0.05 lb and an accuracy of 1%. This versatile instrument can measure both compression and tension, making it suitable for various testing and quality control tasks.
  • Measures both compression and tension for versatile applications.
  • Provides an 11 lb/4.5 kg capacity for a wide range of measurements.
  • Offers a 0.05 lb resolution for precise readings.
  • Ensures reliable results with 1% accuracy.
  • Constructed with a durable plastic housing.
  • Includes a short form certificate of calibration, essential accessories, and a protective case.