Learn More
MSC SPI Analog Compression & Tension Force Gage, 45 lb Capacity, 0.25 lb Resolution, 1% Accuracy
Catalog No. 5000532398
Encompass
This analog compression and tension force gage offers a capacity of 45 lb/22.67 kg with a resolution of 0.25 lb and an accuracy of 1%. Constructed with a durable plastic housing, it is designed to measure both compression and tension.
  • Capacity of 45 lb/22.67 kg
  • Resolution of 0.25 lb
  • Accuracy of 1%
  • Plastic housing material
  • Measures both compression and tension
  • Measuring units in pound and kilogram
  • Includes a short form certificate of calibration, accessories, and a case
  • Available tip types include V-groove, flat, chisel, small hook, and point
Catalog No. 50-005-32398 Supplier MSC Supplier No. 38442125
This analog compression and tension force gage offers a capacity of 45 lb/22.67 kg with a resolution of 0.25 lb and an accuracy of 1%. Constructed with a durable plastic housing, it is designed to measure both compression and tension.
  • Capacity of 45 lb/22.67 kg
  • Resolution of 0.25 lb
  • Accuracy of 1%
  • Plastic housing material
  • Measures both compression and tension
  • Measuring units in pound and kilogram
  • Includes a short form certificate of calibration, accessories, and a case
  • Available tip types include V-groove, flat, chisel, small hook, and point