Learn More
MSC SPI Angle Block, 90 Degree, 4 Inch Long, Steel, 10 Minutes Accuracy
Catalog No. 5000531180
Encompass
This SPI Angle Block is a precision tool designed for various measurement and layout tasks. Made from durable steel, it features a 90-degree angle and a 10-minute accuracy rating, ensuring reliable performance for demanding applications. With a length of 4 inches and a thickness of 1/4 inch, it provides a stable reference for accurate work.
  • Provides a stable 90-degree angle for precise measurement and layout.
  • Constructed from steel for durability and long-lasting use.
  • Offers 10 minutes accuracy for demanding applications.
  • Features a 4 inch length and 1/4 inch thickness for versatile use.
Catalog No. 50-005-31180 Supplier MSC Supplier No. 91099648
This SPI Angle Block is a precision tool designed for various measurement and layout tasks. Made from durable steel, it features a 90-degree angle and a 10-minute accuracy rating, ensuring reliable performance for demanding applications. With a length of 4 inches and a thickness of 1/4 inch, it provides a stable reference for accurate work.
  • Provides a stable 90-degree angle for precise measurement and layout.
  • Constructed from steel for durability and long-lasting use.
  • Offers 10 minutes accuracy for demanding applications.
  • Features a 4 inch length and 1/4 inch thickness for versatile use.