This 3 in x 3 in x 3 in steel precision-ground angle plate is designed for precision layout and inspection setups. Featuring a stepped plate style, machined holes, and an open-end design, it allows for easy clamping and fastening. Made from steel with a precision-ground finish, it boasts an accuracy and squareness of 0.0005 in. It includes 24 holes with a size of 1/4-20, suitable for checking flatness, squaring, and parallels. The stepped design enables efficient clamping and unclamping while in the fixture.
- Machined holes on surface
- Stepped plate style
- Open end design
- Made from steel with a precision-ground finish
- Accuracy of 0.0005 in
- Squareness of 0.0005 in
- Includes 24 holes, size 1/4-20
- Ideal for checking and inspecting flatness, squaring, and parallels
- Stepped design allows for efficient clamping and unclamping of parts