Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC SPI Beveled Straight Edge, 36 in, 0.0008 in Flatness, Spring Steel

Catalog No. 5000529122
Encompass

This SPI Beveled Straight Edge is a precision measuring tool crafted from spring steel, designed for accurate layout and inspection tasks. It features a single beveled edge and offers exceptional flatness and parallelism for reliable performance.

  • Precision tool for accurate layout and inspection.
  • Constructed from durable spring steel.
  • Features a single beveled edge.
  • Edge flatness of 0.0008 inches ensures high accuracy.
  • Parallelism of 0.0016 inches provides reliable measurements.
  • Measures 36 inches in length, 2-13/32 inches in width, and 3/8 inches in thickness.

Catalog No. 50-005-29122 Supplier MSC Supplier No. 02673945
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This SPI Beveled Straight Edge is a precision measuring tool crafted from spring steel, designed for accurate layout and inspection tasks. It features a single beveled edge and offers exceptional flatness and parallelism for reliable performance.

  • Precision tool for accurate layout and inspection.
  • Constructed from durable spring steel.
  • Features a single beveled edge.
  • Edge flatness of 0.0008 inches ensures high accuracy.
  • Parallelism of 0.0016 inches provides reliable measurements.
  • Measures 36 inches in length, 2-13/32 inches in width, and 3/8 inches in thickness.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.