Learn More
MSC SPI Binocular Stereo Microscope, 6.5x-45x, 95 mm Working Distance
Catalog No. 5000530039
Encompass
This stereo microscope offers a versatile magnification range for detailed observation. It features binocular eyepieces and a comfortable working distance, making it suitable for various laboratory and industrial applications.
  • Stereo microscope type
  • Binocular eyepiece type
  • 10x eyepiece magnification
  • 0.65x to 4.5x objective lens magnification
  • 95 mm working distance
  • 6.5x minimum magnification
  • 45x maximum magnification
  • 30 mm to 4.4 mm field of view
Catalog No. 50-005-30039 Supplier MSC Supplier No. 00920496
This stereo microscope offers a versatile magnification range for detailed observation. It features binocular eyepieces and a comfortable working distance, making it suitable for various laboratory and industrial applications.
  • Stereo microscope type
  • Binocular eyepiece type
  • 10x eyepiece magnification
  • 0.65x to 4.5x objective lens magnification
  • 95 mm working distance
  • 6.5x minimum magnification
  • 45x maximum magnification
  • 30 mm to 4.4 mm field of view