Learn More
MSC SPI Swiss Precision Instruments Plug and Pin Gage, 0.3540 in., Black Oxide, 0.000200 in. Tolerance, 1 ea.
Catalog No. 5000514264
Encompass
This Class ZZ minus plug and pin gage is intended for precise measurement and inspection applications. Featuring a 0.3540 in. diameter, this gage is manufactured with a durable black oxide finish.
  • Class ZZ minus accuracy
  • Nominal diameter of 0.3540 in.
  • Black oxide coating for enhanced wear resistance
  • Tolerance of -0.000200 in.
Catalog No. 50-005-14264 Supplier MSC Supplier No. 56247034
This Class ZZ minus plug and pin gage is intended for precise measurement and inspection applications. Featuring a 0.3540 in. diameter, this gage is manufactured with a durable black oxide finish.
  • Class ZZ minus accuracy
  • Nominal diameter of 0.3540 in.
  • Black oxide coating for enhanced wear resistance
  • Tolerance of -0.000200 in.