Learn More
MSC SPI Swiss Precision Instruments Plug Gage, 0.842 in., 0.0002 in. Tolerance, Black Oxide Finish, 1 ea.
Catalog No. 5000547940
Encompass
This SPI Class ZZ plug gage is designed for precision measurements, featuring a 0.842 in. diameter and a 0.0002 in. tolerance. Crafted from hardened drill rod, this gage provides durability and consistent performance in demanding measurement and layout applications.
  • Black oxide coating prevents corrosion and helps identify gage wear
  • Hardened drill rod construction for enhanced durability
  • Overall length of 2 in.
  • Suitable for professional measurement and layout tasks
Catalog No. 50-005-47940 Supplier MSC Supplier No. 56241979
This SPI Class ZZ plug gage is designed for precision measurements, featuring a 0.842 in. diameter and a 0.0002 in. tolerance. Crafted from hardened drill rod, this gage provides durability and consistent performance in demanding measurement and layout applications.
  • Black oxide coating prevents corrosion and helps identify gage wear
  • Hardened drill rod construction for enhanced durability
  • Overall length of 2 in.
  • Suitable for professional measurement and layout tasks