Learn More
MSC SPI Class ZZ Minus Plug Gage, 0.1000 in. Diameter, Black Oxide, 1 ea.
Catalog No. 5000547945
Encompass
This Class ZZ minus plug gage features a diameter of 0.1000 in. and is designed for high-precision measurement applications. It is finished with a black oxide coating for enhanced durability and corrosion resistance.
  • Diameter of 0.1000 in.
  • Class ZZ tolerance of -0.000200 in.
  • Black oxide finish
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Ideal for NoGo inspection tasks
Catalog No. 50-005-47945 Supplier MSC Supplier No. 56244486
This Class ZZ minus plug gage features a diameter of 0.1000 in. and is designed for high-precision measurement applications. It is finished with a black oxide coating for enhanced durability and corrosion resistance.
  • Diameter of 0.1000 in.
  • Class ZZ tolerance of -0.000200 in.
  • Black oxide finish
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Ideal for NoGo inspection tasks