Learn More
MSC SPI Class ZZ Minus Plug Gage, Black Oxide, 0.5700 in. Diameter, 1 ea.
Catalog No. 5000547947
Encompass
This Class ZZ plug and pin gage is designed for precise measurement and inspection applications. Featuring a diameter of 0.5700 in., this tool is constructed for reliable performance in quality control and inspection tasks.
  • Diameter of 0.5700 in.
  • Tolerance of 0.0002 in.
  • Black oxide finish
Catalog No. 50-005-47947 Supplier MSC Supplier No. 56249196
This Class ZZ plug and pin gage is designed for precise measurement and inspection applications. Featuring a diameter of 0.5700 in., this tool is constructed for reliable performance in quality control and inspection tasks.
  • Diameter of 0.5700 in.
  • Tolerance of 0.0002 in.
  • Black oxide finish