Learn More
MSC SPI Class ZZ Minus Plug Gage, 0.6760 in. Diameter, Black Oxide, 1 ea.
Catalog No. 5000547911
Encompass
This Class ZZ plug and pin gage is designed for precision measurements, featuring a 0.6760 in. diameter. The gage is manufactured from hardened drill rod, ensuring durability and consistent performance in demanding measurement and layout applications.
  • Hardened drill rod construction with a hardness of RC 60-62
  • Black oxide coating prevents corrosion and provides visual indication of gage wear
  • Diameter of 0.6760 in. with a tolerance of 0.0002 in.
  • Overall length of 2 in.
Catalog No. 50-005-47911 Supplier MSC Supplier No. 56250251
This Class ZZ plug and pin gage is designed for precision measurements, featuring a 0.6760 in. diameter. The gage is manufactured from hardened drill rod, ensuring durability and consistent performance in demanding measurement and layout applications.
  • Hardened drill rod construction with a hardness of RC 60-62
  • Black oxide coating prevents corrosion and provides visual indication of gage wear
  • Diameter of 0.6760 in. with a tolerance of 0.0002 in.
  • Overall length of 2 in.