Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.9940 in., Black Oxide, 1 ea.
Catalog No. 5000512003
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and inspection applications, including checking micrometers, verniers, and other measuring instruments. Constructed from hardened drill rod with a hardness of RC 60-62, this gage features a durable black oxide finish that provides corrosion resistance and allows for easy identification of gage wear.
  • Diameter of 0.9940 in.
  • Tolerance of +0.000200 in.
  • Overall length of 2 in.
  • Includes certificate of accuracy with NIST traceability
  • Suitable for go inspection and calibration of measuring equipment
Catalog No. 50-005-12003 Supplier MSC Supplier No. 56243512
This Class ZZ Plus plug gage is designed for precise measurement and inspection applications, including checking micrometers, verniers, and other measuring instruments. Constructed from hardened drill rod with a hardness of RC 60-62, this gage features a durable black oxide finish that provides corrosion resistance and allows for easy identification of gage wear.
  • Diameter of 0.9940 in.
  • Tolerance of +0.000200 in.
  • Overall length of 2 in.
  • Includes certificate of accuracy with NIST traceability
  • Suitable for go inspection and calibration of measuring equipment