Learn More
MSC SPI Plug Gage, Black Oxide, 0.8630 in. Diameter, 1 ea.
Catalog No. 5000511989
Encompass
This Class ZZ Plus plug gage is designed for precise diameter measurements and quality control applications. It provides a reliable reference for checking hole sizes and other dimensional specifications in metalworking and manufacturing environments.
  • Features a nominal diameter of 0.8630 in.
  • Constructed with a black oxide finish for enhanced durability and wear resistance.
  • Maintains a tolerance of 0.000200 in. for consistent measurement accuracy.
  • Manufactured in the USA.
Catalog No. 50-005-11989 Supplier MSC Supplier No. 56242183
This Class ZZ Plus plug gage is designed for precise diameter measurements and quality control applications. It provides a reliable reference for checking hole sizes and other dimensional specifications in metalworking and manufacturing environments.
  • Features a nominal diameter of 0.8630 in.
  • Constructed with a black oxide finish for enhanced durability and wear resistance.
  • Maintains a tolerance of 0.000200 in. for consistent measurement accuracy.
  • Manufactured in the USA.