Learn More
MSC SPI Class ZZ Plus Plug Gage, Black Oxide, 0.8220 in. Diameter, 1 ea.
Catalog No. 5000511531
Encompass
This Class ZZ Plus plug gage is designed for high-precision measurement applications. Featuring a 0.8220 in. diameter and a black oxide coating, this tool provides durability and resistance to wear for consistent performance in metalworking and inspection tasks.
  • Diameter: 0.8220 in.
  • Tolerance: +0.0002 in.
  • Finish: Black oxide coating
Catalog No. 50-005-11531 Supplier MSC Supplier No. 56241771
This Class ZZ Plus plug gage is designed for high-precision measurement applications. Featuring a 0.8220 in. diameter and a black oxide coating, this tool provides durability and resistance to wear for consistent performance in metalworking and inspection tasks.
  • Diameter: 0.8220 in.
  • Tolerance: +0.0002 in.
  • Finish: Black oxide coating