Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.9920 in. Diameter, Black Oxide, 1 ea.
Catalog No. 5000511463
Encompass
This Class ZZ plug gage is designed for precise measurement and inspection applications. It features a nominal diameter of 0.9920 in. and is engineered for high accuracy in metalworking and industrial environments.
  • Diameter: 0.9920 in.
  • Tolerance: +0.000200 in.
  • Class: ZZ
  • Finish: Black oxide
Catalog No. 50-005-11463 Supplier MSC Supplier No. 56243496
This Class ZZ plug gage is designed for precise measurement and inspection applications. It features a nominal diameter of 0.9920 in. and is engineered for high accuracy in metalworking and industrial environments.
  • Diameter: 0.9920 in.
  • Tolerance: +0.000200 in.
  • Class: ZZ
  • Finish: Black oxide